Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.5130 in Diameter, Black Oxide
Catalog No. 5000531742
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It features a 0.5130 in diameter and a durable black oxide coating, making it suitable for a variety of industrial and laboratory uses.
  • Designed for precise measurement applications.
  • Features a 0.5130 in diameter.
  • Tolerance of 0.000200 in.
  • Durable black oxide coating.
  • Manufactured in China.
  • Hassle-free guarantee allows for exchange or refund within 30 days of shipment in original packaging.
Catalog No. 50-005-31742 Supplier MSC Supplier No. 56248628
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It features a 0.5130 in diameter and a durable black oxide coating, making it suitable for a variety of industrial and laboratory uses.
  • Designed for precise measurement applications.
  • Features a 0.5130 in diameter.
  • Tolerance of 0.000200 in.
  • Durable black oxide coating.
  • Manufactured in China.
  • Hassle-free guarantee allows for exchange or refund within 30 days of shipment in original packaging.