Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.5130 in Diameter, Steel
Catalog No. 5000530886
Encompass
This product is a Class ZZ Minus Plug and Pin Gage, designed for precise measurement. It features a 0.5130 inch diameter and is constructed from durable steel, ensuring accuracy and longevity in various applications.
  • Measures 0.5130 inch in diameter
  • Constructed from steel for durability
  • Offers a tight tolerance of 0.000200 inch for precise measurements
  • Suitable for various gauging applications
Catalog No. 50-005-30886 Supplier MSC Supplier No. 78377090
This product is a Class ZZ Minus Plug and Pin Gage, designed for precise measurement. It features a 0.5130 inch diameter and is constructed from durable steel, ensuring accuracy and longevity in various applications.
  • Measures 0.5130 inch in diameter
  • Constructed from steel for durability
  • Offers a tight tolerance of 0.000200 inch for precise measurements
  • Suitable for various gauging applications