Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.5310 in Diameter, Black Oxide
Catalog No. 5000511721
Encompass
The SPI Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for accurate diameter verification. It features a 0.5310 in diameter and a durable black oxide finish, suitable for various industrial and laboratory applications.
  • Diameter of measurement: 0.5310 in
  • Tolerance: 0.000200 in
  • Coating/finish: Black oxide
  • Country of origin: China
Catalog No. 50-005-11721 Supplier MSC Supplier No. 56248800
The SPI Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for accurate diameter verification. It features a 0.5310 in diameter and a durable black oxide finish, suitable for various industrial and laboratory applications.
  • Diameter of measurement: 0.5310 in
  • Tolerance: 0.000200 in
  • Coating/finish: Black oxide
  • Country of origin: China