Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.5520 in, 0.000200 in Tolerance
Catalog No. 5000531973
Encompass
This Class ZZ Minus Plug & Pin Gage has a diameter of 0.5520 in. It is made of steel and features a tolerance of 0.000200 in, making it suitable for precise measurement applications.
  • Diameter of measurement: 0.5520 in
  • Tolerance: 0.000200 in
  • Material: Steel
Catalog No. 50-005-31973 Supplier MSC Supplier No. 88955208
This Class ZZ Minus Plug & Pin Gage has a diameter of 0.5520 in. It is made of steel and features a tolerance of 0.000200 in, making it suitable for precise measurement applications.
  • Diameter of measurement: 0.5520 in
  • Tolerance: 0.000200 in
  • Material: Steel