Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.5530 in, Steel
Catalog No. 5000528732
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement. It features a 0.5530 inch diameter, making it suitable for a variety of applications requiring high accuracy.
  • Made from steel
  • Features a 0.5530 inch diameter
  • Provides a 0.000200 inch tolerance
Catalog No. 50-005-28732 Supplier MSC Supplier No. 78377496
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement. It features a 0.5530 inch diameter, making it suitable for a variety of applications requiring high accuracy.
  • Made from steel
  • Features a 0.5530 inch diameter
  • Provides a 0.000200 inch tolerance