Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.5660 in, 0.000200 in
Catalog No. 5000530907
Encompass
Class ZZ Minus Plug & Pin Gages are precision tools designed for accurate measurement of bore diameters. Constructed from durable steel, these gages offer reliable performance with a specified diameter and tolerance, making them suitable for quality control and inspection applications.
  • Designed for accurate measurement of bore diameters.
  • Constructed from durable steel.
  • Features a 0.5660 inch diameter.
  • Maintains a 0.000200 inch tolerance.
  • Ideal for quality control and inspection.
Catalog No. 50-005-30907 Supplier MSC Supplier No. 78377629
Class ZZ Minus Plug & Pin Gages are precision tools designed for accurate measurement of bore diameters. Constructed from durable steel, these gages offer reliable performance with a specified diameter and tolerance, making them suitable for quality control and inspection applications.
  • Designed for accurate measurement of bore diameters.
  • Constructed from durable steel.
  • Features a 0.5660 inch diameter.
  • Maintains a 0.000200 inch tolerance.
  • Ideal for quality control and inspection.