Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.6070 in Diameter, Steel
Catalog No. 5000530793
Encompass
This Class ZZ Minus Plug & Pin Gage features a precise 0.6070 inch diameter and a tolerance of 0.0002 inches. Constructed from durable steel, it is designed for accurate measurement and inspection in various applications.
  • Diameter of 0.6070 inches
  • Tolerance of 0.0002 inches
  • Constructed from steel
  • Ideal for precise measurement and inspection
Catalog No. 50-005-30793 Supplier MSC Supplier No. 78378031
Add to Cart
Add to Cart
This Class ZZ Minus Plug & Pin Gage features a precise 0.6070 inch diameter and a tolerance of 0.0002 inches. Constructed from durable steel, it is designed for accurate measurement and inspection in various applications.
  • Diameter of 0.6070 inches
  • Tolerance of 0.0002 inches
  • Constructed from steel
  • Ideal for precise measurement and inspection