Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.6080 in, Black Oxide
Catalog No. 5000530109
Encompass
This Class ZZ Minus Plug & Pin Gage features a precise 0.6080 inch diameter and a durable black oxide finish. It is designed for accurate measurement applications, including setting micrometers, performing NoGo inspections, and use with other measuring devices.
  • Precise 0.6080 inch diameter
  • Durable black oxide finish
  • Suitable for setting micrometers
  • Ideal for NoGo inspection
  • Compatible with various measuring devices
  • Includes a Certificate of Accuracy with NIST Traceability
Catalog No. 50-005-30109 Supplier MSC Supplier No. 56249576
This Class ZZ Minus Plug & Pin Gage features a precise 0.6080 inch diameter and a durable black oxide finish. It is designed for accurate measurement applications, including setting micrometers, performing NoGo inspections, and use with other measuring devices.
  • Precise 0.6080 inch diameter
  • Durable black oxide finish
  • Suitable for setting micrometers
  • Ideal for NoGo inspection
  • Compatible with various measuring devices
  • Includes a Certificate of Accuracy with NIST Traceability