Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.663 in Diameter, Black Oxide
Catalog No. 5000528728
Encompass
This SPI Class ZZ Minus Plug & Pin Gage features a 0.663 inch diameter and a black oxide finish, making it suitable for a variety of precise measurement and gaging applications.
  • Black oxide finish
  • 0.6630 inch diameter of measurement
  • 0.000200 inch tolerance
  • Designed as a Class ZZ Minus plug and pin gage
Catalog No. 50-005-28728 Supplier MSC Supplier No. 78378593
This SPI Class ZZ Minus Plug & Pin Gage features a 0.663 inch diameter and a black oxide finish, making it suitable for a variety of precise measurement and gaging applications.
  • Black oxide finish
  • 0.6630 inch diameter of measurement
  • 0.000200 inch tolerance
  • Designed as a Class ZZ Minus plug and pin gage