Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.6810" Dia, Black Oxide
Supplier:  MSC 56250301
Encompass
This plug and pin gage is designed for precise measurement with a Class ZZ Minus tolerance. It features a 0.6810 inch diameter and a durable black oxide finish, making it suitable for a variety of inspection and quality control applications.
  • Class ZZ Minus Plug & Pin Gage.
  • 0.6810 inch diameter.
  • Black oxide finish.
  • Designed for high precision measurement.
Catalog No. 50-005-31762