Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.6860 in Dia, Steel
Catalog No. 5000530945
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It features a specific diameter and is made from durable steel, suitable for various industrial and laboratory uses.
  • Diameter of measurement is 0.6860 inches.
  • Tolerance is 0.000200 inches.
  • Constructed from steel.
Catalog No. 50-005-30945 Supplier MSC Supplier No. 78378825
Add to Cart
Add to Cart
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It features a specific diameter and is made from durable steel, suitable for various industrial and laboratory uses.
  • Diameter of measurement is 0.6860 inches.
  • Tolerance is 0.000200 inches.
  • Constructed from steel.