Learn More
MSC SPI Class ZZ Minus Plug and Pin Gage, 0.6920 in Diameter, Steel, 0.0002 in Tolerance
Catalog No. 5000531834
Encompass
This plug and pin gage is designed for precise measurement applications, offering high accuracy for critical dimensional verification. Constructed from durable steel, it ensures reliability and longevity in various industrial and laboratory settings.
  • Constructed from durable steel for longevity.
  • Features a precise diameter of 0.6920 in for accurate gauging.
  • Provides a tight tolerance of 0.0002 in for critical measurements.
  • Designed for use with decimal inch measurement systems.
Catalog No. 50-005-31834 Supplier MSC Supplier No. 56250418
This plug and pin gage is designed for precise measurement applications, offering high accuracy for critical dimensional verification. Constructed from durable steel, it ensures reliability and longevity in various industrial and laboratory settings.
  • Constructed from durable steel for longevity.
  • Features a precise diameter of 0.6920 in for accurate gauging.
  • Provides a tight tolerance of 0.0002 in for critical measurements.
  • Designed for use with decimal inch measurement systems.