Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.6940 in, Black Oxide
Catalog No. 5000531729
Encompass
This Class ZZ Minus Plug & Pin Gage features a precise 0.6940 inch diameter, making it suitable for accurate measurement applications. It is finished with a durable black oxide coating for enhanced longevity and protection.
  • Class ZZ minus plug and pin gage
  • 0.6940 inch diameter of measurement
  • Tolerance of 0.000200 inch
  • Black oxide coating/finish
  • Country of origin: China
Catalog No. 50-005-31729 Supplier MSC Supplier No. 56250434
This Class ZZ Minus Plug & Pin Gage features a precise 0.6940 inch diameter, making it suitable for accurate measurement applications. It is finished with a durable black oxide coating for enhanced longevity and protection.
  • Class ZZ minus plug and pin gage
  • 0.6940 inch diameter of measurement
  • Tolerance of 0.000200 inch
  • Black oxide coating/finish
  • Country of origin: China