Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.7750 in Diameter, 0.000200 in Tolerance, Steel
Catalog No. 5000530190
Encompass
This Class ZZ Minus Plug & Pin Gage features a 0.7750 in diameter and is constructed from steel. It is designed for precise measurements, offering a tight tolerance of 0.000200 in. This gage is suitable for various applications requiring high accuracy in dimensional inspection.
  • Precise 0.7750 in diameter measurement
  • Tight 0.000200 in tolerance for high accuracy
  • Durable steel construction
Catalog No. 50-005-30190 Supplier MSC Supplier No. 78379716
This Class ZZ Minus Plug & Pin Gage features a 0.7750 in diameter and is constructed from steel. It is designed for precise measurements, offering a tight tolerance of 0.000200 in. This gage is suitable for various applications requiring high accuracy in dimensional inspection.
  • Precise 0.7750 in diameter measurement
  • Tight 0.000200 in tolerance for high accuracy
  • Durable steel construction