Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.9100 in Diameter, Black Oxide Finish, 0.000200 in Tolerance
Catalog No. 5000527933
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precision measurement and inspection tasks. Featuring a 0.9100 in diameter and a black oxide finish, it offers accurate gaging for various applications.
  • Provides accurate gaging with a 0.000200 in tolerance.
  • Equipped with a durable black oxide finish.
  • Ideal for setting micrometers, verniers, and other measuring devices.
  • Suitable for NoGo inspection.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Manufactured in China.
Catalog No. 50-005-27933 Supplier MSC Supplier No. 56252612
This Class ZZ Minus Plug & Pin Gage is designed for precision measurement and inspection tasks. Featuring a 0.9100 in diameter and a black oxide finish, it offers accurate gaging for various applications.
  • Provides accurate gaging with a 0.000200 in tolerance.
  • Equipped with a durable black oxide finish.
  • Ideal for setting micrometers, verniers, and other measuring devices.
  • Suitable for NoGo inspection.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Manufactured in China.