Learn More
MSC SPI Class ZZ Minus Plug and Pin Gage, 0.9270 in Diameter, 0.000200 in Tolerance, Steel
Catalog No. 5000528718
Encompass
This Class ZZ Minus Plug and Pin Gage features a 0.9270 inch diameter. Manufactured from steel, it is designed for precise measurement applications. As a made-to-order item, it is not eligible for return or exchange unless found to be defective, and typically has a lead-time of 1 to 2 weeks.
  • Constructed from steel
  • Designed for the decimal inch measurement system
  • Qualifies for free ground shipping within the continental U.S. (excludes Alaska, Hawaii, PO Boxes, and APO/FPO addresses)
  • A minimum order value applies to products in this category
Catalog No. 50-005-28718 Supplier MSC Supplier No. 78381233
This Class ZZ Minus Plug and Pin Gage features a 0.9270 inch diameter. Manufactured from steel, it is designed for precise measurement applications. As a made-to-order item, it is not eligible for return or exchange unless found to be defective, and typically has a lead-time of 1 to 2 weeks.
  • Constructed from steel
  • Designed for the decimal inch measurement system
  • Qualifies for free ground shipping within the continental U.S. (excludes Alaska, Hawaii, PO Boxes, and APO/FPO addresses)
  • A minimum order value applies to products in this category