Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.9730 in, 0.0002 in, Steel
Catalog No. 5000531663
Encompass
The SPI Class ZZ Minus Plug & Pin Gage is a precision instrument designed for accurate measurement. Crafted from durable steel, it ensures reliable performance for various industrial and scientific applications.
  • Provides precise measurements with a diameter of 0.9730 in.
  • Maintains a tight tolerance of 0.0002 in.
  • Constructed from high-quality steel for durability.
  • Features a Class ZZ Minus classification for specific gauging requirements.
Catalog No. 50-005-31663 Supplier MSC Supplier No. 78381696
The SPI Class ZZ Minus Plug & Pin Gage is a precision instrument designed for accurate measurement. Crafted from durable steel, it ensures reliable performance for various industrial and scientific applications.
  • Provides precise measurements with a diameter of 0.9730 in.
  • Maintains a tight tolerance of 0.0002 in.
  • Constructed from high-quality steel for durability.
  • Features a Class ZZ Minus classification for specific gauging requirements.