Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.0120 in, 0.000200 in, Black Oxide
Catalog No. 5000531400
Encompass
This Class ZZ Plus Plug Gage is designed for precision measurement, featuring a 0.0120 in diameter and a strict 0.000200 in tolerance. Its black oxide finish enhances durability and corrosion resistance, making it suitable for a variety of industrial and laboratory applications requiring high accuracy.
  • Class ZZ Plus Plug Gage
  • Precision diameter of 0.0120 in
  • Tight tolerance of 0.000200 in
  • Durable black oxide finish
Catalog No. 50-005-31400 Supplier MSC Supplier No. 84498500
This Class ZZ Plus Plug Gage is designed for precision measurement, featuring a 0.0120 in diameter and a strict 0.000200 in tolerance. Its black oxide finish enhances durability and corrosion resistance, making it suitable for a variety of industrial and laboratory applications requiring high accuracy.
  • Class ZZ Plus Plug Gage
  • Precision diameter of 0.0120 in
  • Tight tolerance of 0.000200 in
  • Durable black oxide finish