Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.0220 in Diameter, 0.000200 in Tolerance, Steel
Catalog No. 5000531521
Encompass
The Class ZZ Plus Plug Gage is a precision measuring tool designed for accurate diameter checks. It features a specific diameter and tolerance, making it suitable for quality control and inspection tasks requiring high accuracy.
  • Precision diameter of 0.0220 inch
  • Tolerance of 0.000200 inch for accurate measurements
  • Constructed from steel for durability
Catalog No. 50-005-31521 Supplier MSC Supplier No. 78382082
The Class ZZ Plus Plug Gage is a precision measuring tool designed for accurate diameter checks. It features a specific diameter and tolerance, making it suitable for quality control and inspection tasks requiring high accuracy.
  • Precision diameter of 0.0220 inch
  • Tolerance of 0.000200 inch for accurate measurements
  • Constructed from steel for durability