Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.0700 in Diameter, 0.000200 in Tolerance, Black Oxide
Catalog No. 5000528932
Encompass
This Class ZZ Plus Plug Gage is designed for precision measurement applications. It features a specific diameter and tolerance, along with a protective black oxide finish, making it suitable for various industrial and laboratory uses.
  • Precisely measures with a Class ZZ Plus Plug Gage.
  • Features a 0.0700 in diameter.
  • Maintains a tight tolerance of 0.000200 in.
  • Coated with black oxide for enhanced durability.
  • Manufactured in China.
Catalog No. 50-005-28932 Supplier MSC Supplier No. 84499094
This Class ZZ Plus Plug Gage is designed for precision measurement applications. It features a specific diameter and tolerance, along with a protective black oxide finish, making it suitable for various industrial and laboratory uses.
  • Precisely measures with a Class ZZ Plus Plug Gage.
  • Features a 0.0700 in diameter.
  • Maintains a tight tolerance of 0.000200 in.
  • Coated with black oxide for enhanced durability.
  • Manufactured in China.