Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.0940 in Dia, 0.000200 in Tolerance
Catalog No. 5000510148
Encompass
This plug gage is designed for precise measurement applications, featuring a steel construction and offering high accuracy for quality control and inspection tasks.
  • Class ZZ plus tolerance
  • 0.0940 in diameter
  • 0.000200 in tolerance
  • Steel material
  • Made in China
Catalog No. 50-005-10148 Supplier MSC Supplier No. 78382793
This plug gage is designed for precise measurement applications, featuring a steel construction and offering high accuracy for quality control and inspection tasks.
  • Class ZZ plus tolerance
  • 0.0940 in diameter
  • 0.000200 in tolerance
  • Steel material
  • Made in China