Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.1070 in Diameter, 0.000200 in Tolerance, Steel
Supplier:  MSC 78382926
Encompass
This Class ZZ Plus plug gage is designed for precise measurement, featuring a 0.1070 inch diameter and a tight tolerance of 0.000200 " It is constructed from steel, ensuring durability and accuracy for various applications.
  • Designed for precise measurement.
  • Features a 0.1070 inch diameter.
  • Provides a tolerance of 0.000200 "
  • Constructed from steel for durability.
Catalog No. 50-005-31551