Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.1210 in Dia, Steel, 0.000200 in Tolerance
Catalog No. 5000528723
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a diameter of 0.1210 inches and a tolerance of 0.000200 inches, ensuring high accuracy. Constructed from steel, it offers durability for reliable use in various industrial and laboratory settings.
  • Diameter of 0.1210 inches
  • Tolerance of 0.000200 inches
  • Constructed from steel
Catalog No. 50-005-28723 Supplier MSC Supplier No. 78383064
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a diameter of 0.1210 inches and a tolerance of 0.000200 inches, ensuring high accuracy. Constructed from steel, it offers durability for reliable use in various industrial and laboratory settings.
  • Diameter of 0.1210 inches
  • Tolerance of 0.000200 inches
  • Constructed from steel