Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.1250 in Dia, 0.000200 in Tolerance
Catalog No. 5000530140
Encompass
This plug gage is designed for precise measurement and inspection applications. It features a specific diameter and tolerance, making it suitable for verifying bore sizes and hole accuracy. Manufactured from durable steel, it offers reliable performance and longevity in various industrial environments.
  • Designed for precise measurement
  • Specific diameter for accurate sizing
  • High tolerance for verifying bore sizes
  • Made from durable steel for longevity
Catalog No. 50-005-30140 Supplier MSC Supplier No. 78383106
This plug gage is designed for precise measurement and inspection applications. It features a specific diameter and tolerance, making it suitable for verifying bore sizes and hole accuracy. Manufactured from durable steel, it offers reliable performance and longevity in various industrial environments.
  • Designed for precise measurement
  • Specific diameter for accurate sizing
  • High tolerance for verifying bore sizes
  • Made from durable steel for longevity