Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.1290 in, 0.000200 in Tolerance
Catalog No. 5000511712
Encompass
This steel Class ZZ Plus plug gage is designed for precise measurement applications. It features a specific diameter and a tight tolerance to ensure accuracy in inspection.
  • Designed for precise measurement.
  • Diameter of 0.1290 "
  • Tolerance of 0.000200 "
  • Constructed from steel.
Catalog No. 50-005-11712 Supplier MSC Supplier No. 78383148
This steel Class ZZ Plus plug gage is designed for precise measurement applications. It features a specific diameter and a tight tolerance to ensure accuracy in inspection.
  • Designed for precise measurement.
  • Diameter of 0.1290 "
  • Tolerance of 0.000200 "
  • Constructed from steel.