Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.1700 Dia, 0.0002 Tol, Black Oxide
Catalog No. 5000530403
Encompass
This Class ZZ Plus Plug Gage is engineered for highly accurate measurement tasks. It provides a precise 0.1700 inch diameter and features a tight 0.0002 inch tolerance, ensuring reliability in quality control and inspection processes. The durable black oxide finish offers enhanced protection and longevity.
  • Features a 0.1700 inch diameter measurement
  • Offers a tight 0.0002 inch tolerance
  • Equipped with a protective black oxide finish
  • Ideal for precision measurement applications
Catalog No. 50-005-30403 Supplier MSC Supplier No. 84500099
This Class ZZ Plus Plug Gage is engineered for highly accurate measurement tasks. It provides a precise 0.1700 inch diameter and features a tight 0.0002 inch tolerance, ensuring reliability in quality control and inspection processes. The durable black oxide finish offers enhanced protection and longevity.
  • Features a 0.1700 inch diameter measurement
  • Offers a tight 0.0002 inch tolerance
  • Equipped with a protective black oxide finish
  • Ideal for precision measurement applications