Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.1990 in, 0.000200 in Tolerance
Supplier:  MSC 84500388
Encompass
This Class ZZ Plus Plug Gage is engineered for precise measurement applications. It features a 0.1990 in diameter with a tight tolerance of 0.000200 in, and a black oxide finish for durability.
  • Designed for precise measurement applications.
  • Features a 0.1990 in diameter.
  • Offers a tight 0.000200 in tolerance.
  • Equipped with a black oxide finish for durability.
Catalog No. 50-005-31305