Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.2150 in, 0.000200 in Tolerance, Steel
Catalog No. 5000529010
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a diameter of 0.2150 inches and a tolerance of 0.000200 inches, providing accuracy for various tasks. Made from durable steel, it ensures long-lasting performance.
  • Diameter of measurement: 0.2150 in
  • Tolerance: 0.000200 in
  • Material: Steel
  • Manufactured in China
Catalog No. 50-005-29010 Supplier MSC Supplier No. 78384005
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a diameter of 0.2150 inches and a tolerance of 0.000200 inches, providing accuracy for various tasks. Made from durable steel, it ensures long-lasting performance.
  • Diameter of measurement: 0.2150 in
  • Tolerance: 0.000200 in
  • Material: Steel
  • Manufactured in China