Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.2410 in Dia, 0.0002 in Tolerance, Steel
Catalog No. 5000530770
Encompass
This plug gage is designed for precise measurement and calibration in various industrial and scientific applications. It ensures accurate verification of hole diameters and other critical dimensions.
  • Class ZZ Plus for high precision
  • 0.2410 in diameter for specific measurements
  • 0.0002 in tolerance for accuracy
  • Constructed from durable steel for longevity
  • Ideal for quality control and inspection tasks
Catalog No. 50-005-30770 Supplier MSC Supplier No. 78384260
This plug gage is designed for precise measurement and calibration in various industrial and scientific applications. It ensures accurate verification of hole diameters and other critical dimensions.
  • Class ZZ Plus for high precision
  • 0.2410 in diameter for specific measurements
  • 0.0002 in tolerance for accuracy
  • Constructed from durable steel for longevity
  • Ideal for quality control and inspection tasks