Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.2440 in Diameter, 0.000200 in Tolerance
Catalog No. 5000527348
Encompass
This Class ZZ Plus Plug Gage features a diameter of 0.2440 in and a tolerance of 0.000200 in. It is finished with a black oxide coating.
  • Diameter of measurement: 0.2440 in
  • Tolerance: 0.000200 in
  • Black oxide coating
Catalog No. 50-005-27348 Supplier MSC Supplier No. 84500834
Add to Cart
Add to Cart
This Class ZZ Plus Plug Gage features a diameter of 0.2440 in and a tolerance of 0.000200 in. It is finished with a black oxide coating.
  • Diameter of measurement: 0.2440 in
  • Tolerance: 0.000200 in
  • Black oxide coating