Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.2590 in Diameter, 0.000200 in Tolerance
Catalog No. 5000528835
Encompass
This Class ZZ Plus Plug Gage is designed for accurate measurement, featuring a precise diameter and tolerance for quality control and calibration applications. Made from durable steel, it ensures reliable performance in industrial and laboratory environments.
  • Diameter of 0.2590 in
  • Tolerance of 0.000200 in
  • Constructed from steel for durability
  • Manufactured in China
Catalog No. 50-005-28835 Supplier MSC Supplier No. 78384443
This Class ZZ Plus Plug Gage is designed for accurate measurement, featuring a precise diameter and tolerance for quality control and calibration applications. Made from durable steel, it ensures reliable performance in industrial and laboratory environments.
  • Diameter of 0.2590 in
  • Tolerance of 0.000200 in
  • Constructed from steel for durability
  • Manufactured in China