Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.2930 in, 0.000200 in Tolerance, Black oxide
Catalog No. 5000527228
Encompass
This Class ZZ Plus Plug Gage features a 0.2930 inch diameter and a 0.000200 inch tolerance, with a black oxide finish. It is designed for 'Go' inspection and for setting micrometers, verniers, and other measuring devices. A Certificate of Accuracy with NIST Traceability is included, and the gage meets ASME B89.1 standards.
  • Used for 'Go' inspection
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • 0.2930 in diameter
  • 0.000200 in tolerance
  • Black oxide finish
  • Country of origin: China
Catalog No. 50-005-27228 Supplier MSC Supplier No. 84501329
This Class ZZ Plus Plug Gage features a 0.2930 inch diameter and a 0.000200 inch tolerance, with a black oxide finish. It is designed for 'Go' inspection and for setting micrometers, verniers, and other measuring devices. A Certificate of Accuracy with NIST Traceability is included, and the gage meets ASME B89.1 standards.
  • Used for 'Go' inspection
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • 0.2930 in diameter
  • 0.000200 in tolerance
  • Black oxide finish
  • Country of origin: China