Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.3110 in, 0.000200 in, Black Oxide
Catalog No. 5000527221
Encompass
This Class ZZ Plus plug gage is designed for precise measurement applications. It features a specific diameter and tolerance, along with a protective finish, making it suitable for various quality control and inspection tasks.
  • Designed for precise measurements with a diameter of 0.3110 inches (7.90 mm).
  • Features a tolerance of 0.000200 inches (0.0051 mm) for accuracy.
  • Enhanced with a durable black oxide coating.
Catalog No. 50-005-27221 Supplier MSC Supplier No. 84501519
This Class ZZ Plus plug gage is designed for precise measurement applications. It features a specific diameter and tolerance, along with a protective finish, making it suitable for various quality control and inspection tasks.
  • Designed for precise measurements with a diameter of 0.3110 inches (7.90 mm).
  • Features a tolerance of 0.000200 inches (0.0051 mm) for accuracy.
  • Enhanced with a durable black oxide coating.