Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.3340 in Dia, 0.0002 in Tolerance, Black Oxide
Catalog No. 5000510775
Encompass
This Class ZZ Plus Plug Gage has a diameter of 0.3340 in and a tolerance of 0.0002 in. It features a durable black oxide coating, making it suitable for precise measurement applications.
  • Class ZZ plus plug gage
  • 0.3340 in diameter
  • 0.0002 in tolerance
  • Black oxide coating
Catalog No. 50-005-10775 Supplier MSC Supplier No. 84501741
This Class ZZ Plus Plug Gage has a diameter of 0.3340 in and a tolerance of 0.0002 in. It features a durable black oxide coating, making it suitable for precise measurement applications.
  • Class ZZ plus plug gage
  • 0.3340 in diameter
  • 0.0002 in tolerance
  • Black oxide coating