Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.3510 in Diameter, 0.000200 in Tolerance, Steel
Catalog No. 5000530314
Encompass
This Class ZZ Plus plug gage is designed for precise measurement applications, offering high accuracy for quality control and inspection tasks. It features a specific diameter and tolerance, manufactured from durable steel.
  • Engineered for precise measurement applications.
  • Features a 0.3510 in diameter.
  • Provides a tolerance of 0.000200 in.
  • Constructed from durable steel for longevity.
  • Suitable for quality control and inspection tasks.
Catalog No. 50-005-30314 Supplier MSC Supplier No. 78385366
This Class ZZ Plus plug gage is designed for precise measurement applications, offering high accuracy for quality control and inspection tasks. It features a specific diameter and tolerance, manufactured from durable steel.
  • Engineered for precise measurement applications.
  • Features a 0.3510 in diameter.
  • Provides a tolerance of 0.000200 in.
  • Constructed from durable steel for longevity.
  • Suitable for quality control and inspection tasks.