Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.3770 in, 0.000200 in Tolerance
Catalog No. 5000531507
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a 0.3770 in diameter and a tolerance of 0.000200 in, with a durable black oxide finish.
  • Precise measurement of 0.3770 in diameter
  • Tight tolerance of 0.000200 in for accuracy
  • Durable black oxide finish for enhanced longevity
  • Ideal for quality control and manufacturing inspections
Catalog No. 50-005-31507 Supplier MSC Supplier No. 84502178
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a 0.3770 in diameter and a tolerance of 0.000200 in, with a durable black oxide finish.
  • Precise measurement of 0.3770 in diameter
  • Tight tolerance of 0.000200 in for accuracy
  • Durable black oxide finish for enhanced longevity
  • Ideal for quality control and manufacturing inspections