Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.3820 in Diameter, 0.000200 in Tolerance, Black Oxide
Catalog No. 5000527222
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement, featuring a diameter of 0.3820 inches and a tight tolerance of 0.000200 inches. It is finished with a durable black oxide coating.
  • Accurate diameter of 0.3820 inches for precise measurements.
  • Tight tolerance of 0.000200 inches ensures high accuracy.
  • Durable black oxide finish for extended product life.
Catalog No. 50-005-27222 Supplier MSC Supplier No. 84502228
This Class ZZ Plus Plug Gage is designed for precise measurement, featuring a diameter of 0.3820 inches and a tight tolerance of 0.000200 inches. It is finished with a durable black oxide coating.
  • Accurate diameter of 0.3820 inches for precise measurements.
  • Tight tolerance of 0.000200 inches ensures high accuracy.
  • Durable black oxide finish for extended product life.