Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.4090 in Diameter, Steel
Catalog No. 5000530932
Encompass
This plug gage is a Class ZZ Plus model, designed for precise measurements. It is manufactured from steel and features a specific diameter and tolerance for accurate gauging.
  • Measures 0.4090 in diameter
  • Provides a tolerance of 0.000200 in
Catalog No. 50-005-30932 Supplier MSC Supplier No. 78385945
This plug gage is a Class ZZ Plus model, designed for precise measurements. It is manufactured from steel and features a specific diameter and tolerance for accurate gauging.
  • Measures 0.4090 in diameter
  • Provides a tolerance of 0.000200 in