Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.4220 in, Black Oxide
Catalog No. 5000528622
Encompass
This SPI Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a 0.4220 in diameter and a black oxide finish, providing durability and accuracy for various quality control and inspection tasks.
  • Features a 0.4220 in diameter.
  • Has a tolerance of 0.000200 in.
  • Includes a black oxide coating.
  • Manufactured in China.
  • Suitable for quality control and inspection tasks.
Catalog No. 50-005-28622 Supplier MSC Supplier No. 50741586
This SPI Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a 0.4220 in diameter and a black oxide finish, providing durability and accuracy for various quality control and inspection tasks.
  • Features a 0.4220 in diameter.
  • Has a tolerance of 0.000200 in.
  • Includes a black oxide coating.
  • Manufactured in China.
  • Suitable for quality control and inspection tasks.