Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.4430 in Dia, 0.000200 in Tolerance, Black Oxide
Catalog No. 5000528950
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a 0.4430 inch diameter and a tight tolerance of 0.000200 inch, ensuring accuracy for critical inspections. The black oxide finish provides enhanced durability and corrosion resistance, making it a reliable tool for quality control.
  • Features a 0.4430 inch diameter
  • Offers a tolerance of 0.000200 inch
  • Equipped with a black oxide coating for enhanced durability and corrosion resistance
  • Ideal for precise measurement applications
Catalog No. 50-005-28950 Supplier MSC Supplier No. 84502830
Add to Cart
Add to Cart
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a 0.4430 inch diameter and a tight tolerance of 0.000200 inch, ensuring accuracy for critical inspections. The black oxide finish provides enhanced durability and corrosion resistance, making it a reliable tool for quality control.
  • Features a 0.4430 inch diameter
  • Offers a tolerance of 0.000200 inch
  • Equipped with a black oxide coating for enhanced durability and corrosion resistance
  • Ideal for precise measurement applications