Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.4530 in Diameter, 0.000200 in Tolerance, Steel
Catalog No. 5000530903
Encompass
This Class ZZ Plus Plug Gage features a diameter of 0.4530 in and a tolerance of 0.000200 in. It is constructed from durable steel, suitable for precision measurement applications.
  • Diameter of 0.4530 in
  • Tolerance of 0.000200 in
  • Made of steel
  • Manufactured in China
Catalog No. 50-005-30903 Supplier MSC Supplier No. 78386380
This Class ZZ Plus Plug Gage features a diameter of 0.4530 in and a tolerance of 0.000200 in. It is constructed from durable steel, suitable for precision measurement applications.
  • Diameter of 0.4530 in
  • Tolerance of 0.000200 in
  • Made of steel
  • Manufactured in China