Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.4560 Dia, 0.000200 Tolerance, Black Oxide
Catalog No. 5000527604
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a 0.4560 inch diameter with a tight tolerance of 0.000200 inch, and is finished with black oxide for enhanced durability.
  • Class ZZ plus plug gage
  • Diameter of measurement 0.4560 in
  • Tolerance of 0.000200 in
  • Black oxide coating/finish
Catalog No. 50-005-27604 Supplier MSC Supplier No. 84502962
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a 0.4560 inch diameter with a tight tolerance of 0.000200 inch, and is finished with black oxide for enhanced durability.
  • Class ZZ plus plug gage
  • Diameter of measurement 0.4560 in
  • Tolerance of 0.000200 in
  • Black oxide coating/finish