Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.4590 in Dia, 0.0002 in Tolerance, Black Oxide
Catalog No. 5000527230
Encompass
This Class ZZ Plus Plug Gage features a 0.4590 in diameter and a 0.0002 in tolerance. It is finished with black oxide, making it suitable for precise measurement applications.
  • Diameter of measurement: 0.4590 in
  • Tolerance: 0.0002 in
  • Black oxide coating
Catalog No. 50-005-27230 Supplier MSC Supplier No. 84502996
This Class ZZ Plus Plug Gage features a 0.4590 in diameter and a 0.0002 in tolerance. It is finished with black oxide, making it suitable for precise measurement applications.
  • Diameter of measurement: 0.4590 in
  • Tolerance: 0.0002 in
  • Black oxide coating