Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.4860 in Dia, 0.000200 in Tolerance, Black Oxide
Catalog No. 5000510697
Encompass
This Class ZZ Plus plug gage is designed for precise measurement and inspection applications. Featuring a 0.4860 in diameter and a black oxide finish, it offers a tolerance of 0.000200 in for accurate gauging.
  • Class ZZ Plus plug gage
  • 0.4860 in diameter
  • 0.000200 in tolerance
  • Black oxide finish
  • Suitable for precise measurement and inspection
Catalog No. 50-005-10697 Supplier MSC Supplier No. 84503267
This Class ZZ Plus plug gage is designed for precise measurement and inspection applications. Featuring a 0.4860 in diameter and a black oxide finish, it offers a tolerance of 0.000200 in for accurate gauging.
  • Class ZZ Plus plug gage
  • 0.4860 in diameter
  • 0.000200 in tolerance
  • Black oxide finish
  • Suitable for precise measurement and inspection