Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.4900 in Diameter, 0.000200 in Tolerance, Black Oxide
Catalog No. 5000531206
Encompass
This plug gage is designed for precise measurement applications. It features a 0.4900 inch diameter and a tolerance of 0.000200 inch, with a durable black oxide finish.
  • Diameter of measurement: 0.4900 in
  • Tolerance: 0.000200 in
  • Black oxide coating/finish
Catalog No. 50-005-31206 Supplier MSC Supplier No. 84503309
This plug gage is designed for precise measurement applications. It features a 0.4900 inch diameter and a tolerance of 0.000200 inch, with a durable black oxide finish.
  • Diameter of measurement: 0.4900 in
  • Tolerance: 0.000200 in
  • Black oxide coating/finish