Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.5000 in Diameter, 0.000200 in Tolerance, Black Oxide
Catalog No. 5000527576
Encompass
This Class ZZ Plus Plug Gage is designed for precision measurement applications. It features a 0.5000 inch diameter and a 0.000200 inch tolerance, with a durable black oxide finish.
  • Features a 0.5000 inch diameter.
  • Offers a 0.000200 inch tolerance for precise measurements.
  • Finished with black oxide for enhanced durability.
Catalog No. 50-005-27576 Supplier MSC Supplier No. 84503408
This Class ZZ Plus Plug Gage is designed for precision measurement applications. It features a 0.5000 inch diameter and a 0.000200 inch tolerance, with a durable black oxide finish.
  • Features a 0.5000 inch diameter.
  • Offers a 0.000200 inch tolerance for precise measurements.
  • Finished with black oxide for enhanced durability.