Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.5020 in Diameter, Black Oxide, 0.000200 in Tolerance
Catalog No. 5000528948
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a 0.5020 inch diameter and a black oxide coating, providing durability and accuracy for various industrial uses.
  • Provides precise measurement for quality control
  • Features a durable black oxide finish
  • Offers a 0.5020 inch diameter for specific gauging needs
  • Ensures high accuracy with a 0.000200 inch tolerance
Catalog No. 50-005-28948 Supplier MSC Supplier No. 84503424
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a 0.5020 inch diameter and a black oxide coating, providing durability and accuracy for various industrial uses.
  • Provides precise measurement for quality control
  • Features a durable black oxide finish
  • Offers a 0.5020 inch diameter for specific gauging needs
  • Ensures high accuracy with a 0.000200 inch tolerance