Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.5050 in Diameter, 0.0002 in Tolerance, Steel
Catalog No. 5000530828
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement applications, offering exceptional accuracy with a diameter of 0.5050 inches and a tight tolerance of 0.0002 inches. Constructed from durable steel, it ensures reliability and longevity in various industrial environments.
  • Precise diameter for accurate measurements.
  • Tight tolerance for high-precision gauging.
  • Durable steel construction for extended lifespan.
  • Suitable for a wide range of inspection tasks.
Catalog No. 50-005-30828 Supplier MSC Supplier No. 78386901
This Class ZZ Plus Plug Gage is designed for precise measurement applications, offering exceptional accuracy with a diameter of 0.5050 inches and a tight tolerance of 0.0002 inches. Constructed from durable steel, it ensures reliability and longevity in various industrial environments.
  • Precise diameter for accurate measurements.
  • Tight tolerance for high-precision gauging.
  • Durable steel construction for extended lifespan.
  • Suitable for a wide range of inspection tasks.