Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.525 in, 0.0002 in, Black Oxide Finish
Catalog No. 5000527225
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement applications, offering a specific diameter and tight tolerance to ensure accuracy. It also features a black oxide finish for enhanced durability and corrosion resistance.
  • Diameter of measurement: 0.525 in
  • Tolerance: 0.0002 in
  • Black oxide finish
Catalog No. 50-005-27225 Supplier MSC Supplier No. 84503655
This Class ZZ Plus Plug Gage is designed for precise measurement applications, offering a specific diameter and tight tolerance to ensure accuracy. It also features a black oxide finish for enhanced durability and corrosion resistance.
  • Diameter of measurement: 0.525 in
  • Tolerance: 0.0002 in
  • Black oxide finish