Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.5380 inch Diameter, Steel
Catalog No. 5000527266
Encompass
The SPI Class ZZ Plus Plug Gage is a steel plug gage designed for precise measurement and inspection. It features a 0.5380 inch diameter and a tolerance of 0.000200 "
  • Designed for precise measurement and inspection.
  • Constructed from durable steel.
  • Features a 0.5380 inch diameter.
  • Offers a 0.000200 inch tolerance.
  • Manufactured in China.
Catalog No. 50-005-27266 Supplier MSC Supplier No. 78387230
Add to Cart
Add to Cart
The SPI Class ZZ Plus Plug Gage is a steel plug gage designed for precise measurement and inspection. It features a 0.5380 inch diameter and a tolerance of 0.000200 "
  • Designed for precise measurement and inspection.
  • Constructed from durable steel.
  • Features a 0.5380 inch diameter.
  • Offers a 0.000200 inch tolerance.
  • Manufactured in China.